Order Entry
Denmark
ContactUsLinkComponent
Agarose for electrophoresis low electroendosmosis (EEO) (Mr <= 0.10)
Agarose for electrophoresis low electroendosmosis (EEO) (Mr <= 0.10)
Certificates
undefined
Agarose for electrophoresis low electroendosmosis (EEO) (Mr <= 0.10)
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 %
  • Gel strength:
    ≥800 g/cm² (1% gel)
  • Gel point:
    34 to 45°C
  • Electroendosmosis:
    ≤0.12 (-Mr)
  • Sulfate (SO4):
    ≤0.3 %