Order Entry
Denmark
ContactUsLinkComponent
Gentamicin sulphate 50 mg/ml in aqueous solution, sterile filtered, Millipore®
Gentamicin sulphate 50 mg/ml in aqueous solution, sterile filtered, Millipore®
Catalog # 345815-20ML
CAS Number:  
Gentamicin sulphate 50 mg/ml in aqueous solution, sterile filtered, Millipore®
Catalog # 345815-20ML
CAS Number:  

Specifications

  • Pk:
    20 mL
  • Seq:
    2305

Specifications

About this item

 :
CAS Number:1405-41-0
MDL Number:MFCD00270181