Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose, GPR RECTAPUR®

D-(-)-Fructose, GPR RECTAPUR®

Plastic bottle for solids, 1 kg
Catalog # 24975.294
Supplier:   VWR Chemicals
D-(-)-Fructose, GPR RECTAPUR®
Catalog # 24975.294
Supplier:  VWR Chemicals
CAS Number:  57-48-7
D-(-)-Fructose, GPR RECTAPUR®
D-(-)-Fructose, GPR RECTAPUR®

Plastic bottle for solids, 1 kg

CAS Number:  57-48-7
Price
593,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Specific optical rotation (10 %; water):
    -94 to -90 °
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    Max. 0.5 %
  • Cl (Chloride):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 100 ppm

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.