Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose ≥99%, high purity

D-(-)-Fructose ≥99%, high purity

D-(-)-Fructose ≥99%, high purity
Supplier:  VWR Chemicals
Certificates
D-(-)-Fructose ≥99%, high purity

2 Options Available Below

Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications for Product Family

Specification Test Results

  • Arsenic:
    <= 0.0001%
  • Calcium & Magnesium:
    <= 0.005%
  • Chloride:
    <= 0.018%
  • Heavy Metals (as Pb):
    <= 0.0005%
  • Loss on Drying:
    <= 0.5%
  • Purity:
    >= 99%
  • Residue on Ignition:
    <= 0.5%
  • Sulphate:
    <= 0.025%
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.