Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose ≥99%

D-(-)-Fructose ≥99%

D-(-)-Fructose ≥99%
Certificates
D-(-)-Fructose ≥99%
D-(-)-Fructose ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 57-48-7 Molecular Formula: C6H12O6 Formula Weight: 180.16 Odour: Odourless Melting Point: ca 105-110° Beilstein Reference: 1239004 Merck Reference: 14,4273 Optical Rotation: -89° (c=2 in water) MDL No.: MFCD00148910

Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,4273
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:∼ 105…110 °C
  • Density:1.59
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 409,00 kr
    409,00 kr
    1 * 250 g
       409,00 kr

  • Glass bottle for solids
    1.000 g
    Log in for availability
    Log in for availability
    1 * 1.000 g
    1 * 1.000 g - 892,00 kr
    892,00 kr
    1 * 1.000 g
       892,00 kr

  • Glass bottle for solids
    5.000 g
    Log in for availability
    Log in for availability
    1 * 5.000 g
    1 * 5.000 g - 3.100,00 kr
    3.100,00 kr
    1 * 5.000 g
       3.100,00 kr