Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose 99%

D-(-)-Fructose 99%

D-(-)-Fructose 99%
Certificates
D-(-)-Fructose 99%

4 Options Available Below

Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:103…105 °C (dec.)
  • Density:1,59 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-333-3

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥98.5 %
  • Loss on drying:
    ≤0.5 % (70°C, 4 hrs) (Vacuum)
  • Sulfated ash:
    ≤0.1 %
  • Additional info:
    5-Hydroxymethylfurfural and rel. compounds: Passes
  • Appearance of solution:
    Passes test
  • Specific optical rotation:
    -91.0° to -93.5° (20°C, 589 nm) (c=10, H2O/NH3) (After 30 min, On dry substance)
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Foreign sugars:
    Passes test
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.