D-(+)-Trehalose, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:99-20-7
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006628
:342.3 g/mol
Merck Index:12,09713
Melting Pt:203 °C
Density:1.53 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:202-739-6
Related Information
Certificates
Search by Lot Numbermsds_108216
SDS
ProductPage_108216 ProductPage_108216
Product specification
Frequently Bought Together