Jump to:
About this item
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Melting Pt:83 °C
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:200-075-1
3 Options Available Below
Product Details & Documents
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Melting Pt:83 °C
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:200-075-1



