D-(+)-Glucose monohydrate
Supplier: Thermo Fisher Scientific
Jump to:
About this item
CAS No.: 14431-43-7 Molecular Formula: C6H12O6.H2O Formula Weight: 198.18 (180.16anhy) Physical Form: Cryst powder Appearance: White Melting Point: ca 83° MDL No.: MFCD00149450
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Melting Pt:∼ 83 °C
- Storage Temperature:Ambient
- EINECS:200-075-1
2 Options Available Below
Product Details & Documents
CAS No.: 14431-43-7 Molecular Formula: C6H12O6.H2O Formula Weight: 198.18 (180.16anhy) Physical Form: Cryst powder Appearance: White Melting Point: ca 83° MDL No.: MFCD00149450
Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
- CAS Number:14431-43-7
- Formula:C₆H₁₂O₆·H₂O
- MDL Number:MFCD00149450
- Molecular Weight:198.17 g/mol
- Melting Pt:∼ 83 °C
- Storage Temperature:Ambient
- EINECS:200-075-1



