D-(+)-Maltose monohydrate, Millipore®
Leverandør: Merck
Certifikater
Om denne vare
CAS nummer:6363-53-7
:
Formel:C₁₂H₂₂O₁₁·1H₂O
MDL nummer:MFCD00149343
:360.32 g/mol
Merck Index:13,05736
Smeltende Pt:160…165 °C
:M-Clarity™ quality level = MQ100
Opbevaringstemperatur:Ambient
EINECS:200-716-5
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Relateret information
Frequently Bought Together