Order Entry
Denmark
ContactUsLinkComponent
 
D-(+)-Maltose monohydrate ≥95%, white powder
 
undefined
D-(+)-Maltose monohydrate ≥95%, white powder

 

  • Identity Test:
    Passes
  • Purity:
    ≥95%
  • FTIR:
    Conforms to Standard
  • Loss on Drying:
    ≤6%
  • Optical Rotation:
    130.0 ± 2.0 (c=4, water)
  • Solubility (10% aq soln):
    Clear and Colorless

 

Frequently Bought Together