Order Entry
Denmark
ContactUsLinkComponent
Pyridine-[D₅]
Flammable
Irritant
Pyridine-[D₅]
Catalog # EHERC16646100
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Irritant
Pyridine-[D₅]
Catalog # EHERC16646100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :84.06 g/mol
Boiling Pt:114.4 °C (1013 hPa)
Melting Pt:–41 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:230-720-2
UN:1282
ADR:3,II
 :GHS02
 :GHS07
CAS Number:7291-22-7
Formula:C₅D₅N
MDL Number:MFCD00044639