Order Entry
Denmark
ContactUsLinkComponent
 
Dimethyl sulphoxide-[D₆] 100.0 atom % D, > 99.96 atom % D for NMR spectroscopy
  ACRO169020250
 :  
undefined
Dimethyl sulphoxide-[D₆] 100.0 atom % D, > 99.96 atom % D for NMR spectroscopy
  ACRO169020250
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.95 %

 

 

 :
 :84.09 g/mol
 :189 °C (5 mmHg)
 :20.2 °C
 :1.19 g/cm³
 :88 °C (closed cup)
 :Ambient
 :218-617-0
 :2206-27-1
 :D₃CS(O)CD₃
 :MFCD00002090