Deuterium oxide 99.8 atom % D for NMR spectroscopy
Catalog # ACRO351430075
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:7,5 mL
- Seq:2065
- Pk Info:10 x 0.75 ml
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- D-Enrichment:≥99.75 %
- Tritium (T):≤370 Bq/g
Specification Test Results
About this item
:
:20 g/mol
Merck Index:12,02984
Boiling Pt:101.3 °C (1013 hPa)
Melting Pt:3.8 °C
Density:1.105 g/cm³
Storage Temperature:Refrigerator
EINECS:232-148-9
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636