Order Entry
Denmark
Orders LinkContactUsLinkComponent
Benzene-[D₆]

Benzene-[D₆]

Flammable
Health
Marketsource
Supplier:   Roth Carl
Benzene-[D₆]
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Benzene-[²H₆]
  • CAS Number:1076-43-3
  • Formula:C₆D₆
  • MDL Number:MFCD00003010
  • Molecular Weight:84.07 g/mol
  • Boiling Pt:79 °C (1013 hPa)
  • Melting Pt:6.7 °C
  • Density:0.95 g/cm³ (20 °C)
  • Flash Pt:–11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:214-061-8
  • UN:1114
  • ADR:3,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 671,34 kr
    671,34 kr
    1 * 10 mL
       671,34 kr

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 2.680,88 kr
    2.680,88 kr
    1 * 50 mL
       2.680,88 kr

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.