Order Entry
Denmark
ContactUsLinkComponent
Benzene-[D₆] (99.5% D) + 0.03% v/v TMS
Flammable
Health
Benzene-[D₆] (99.5% D) + 0.03% v/v TMS
Catalog # CAMBDLM-1TB-10
CAS Number:  
Flammable
Health
Benzene-[D₆] (99.5% D) + 0.03% v/v TMS
Catalog # CAMBDLM-1TB-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :84.07 g/mol
Boiling Pt:79 °C (1013 hPa)
Melting Pt:6.7 °C
Density:0.95 g/cm³ (20 °C)
Flash Pt:–11 °C (closed cup)
Storage Temperature:Ambient
EINECS:214-061-8
UN:1114
ADR:3,II
 :GHS02
 :GHS08
CAS Number:1076-43-3
Formula:C₆D₆
MDL Number:MFCD00003010