

Trifluoroacetic acid-[D₁] (99.5% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.13363.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:115.02 g/mol
Boiling Pt:75 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.5 g/cm³ (20 °C)
EINECS:209-961-2
UN:2699
ADR:8,
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
CAS Number:599-00-8
Formula:F₃CCOOD
MDL Number:MFCD00044563