

Trifluoroacetic acid-[D₁] (99.5% D)
Catalog # CAMBDLM-46-10
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:115.02 g/mol
Boiling Pt:75 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.5 g/cm³ (20 °C)
EINECS:209-961-2
UN:2699
ADR:8,I
:GHS05
:GHS07
CAS Number:599-00-8
Formula:F₃CCOOD
MDL Number:MFCD00044563