Chloroform-[D₁] (99.8% D) for NMR spectroscopy
Catalog # CAMBDLM-7-100
Supplier: CAMBRIDGE ISOTOPE LABS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D
:
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64.1 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
:GHS08
:GHS07
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827