Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beeswax

Beeswax

Marketsource
Supplier:   Roth Carl
Beeswax
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Wax, bees
  • CAS Number:8012-89-3
  • MDL Number:MFCD00132754
  • Melting Pt:∼64 °C
  • Density:0.95 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-383-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.