Beeswax, yellow, TECHNICAL
Plastic bottle for solids, 500 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Wax, bees
- CAS Number:8012-89-3
- MDL Number:MFCD00132754
- Melting Pt:∼64 °C
- Density:0.95 g/cm³ (20 °C)
- EINECS:232-383-7
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
P20_VWRC22842.268_SPEC_EN.pdf spec_VWRC22842.268 spec_VWRC22842.268
Product specification
Specifications
Specification Test Results
- Melting point:61 - 65 °C
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



