Order Entry
Denmark
ContactUsLinkComponent
Petroleum spirit, 80…100 °C, EMSURE® for analysis, Supelco®
Flammable
Health
Environment
Petroleum spirit, 80…100 °C, EMSURE® for analysis, Supelco®
Catalog # 1.01777.1000
Supplier:  Merck
CAS Number:  
Flammable
Health
Environment
Petroleum spirit, 80…100 °C, EMSURE® for analysis, Supelco®
Catalog # 1.01777.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (25 °C) insoluble
  • Boiling Point:
    80 - 100 °C (1013 hPa)
  • Vapor Pressure:
    60 hPa (20 °C)
  • Explosion Limit:
    1.0 - 6.5 %(V)
  • Density:
    0.69 g/cm3 (20 °C)
  • Ignition Point:
    250 °C
  • Viscosity kinematic:
    0.6 mm2/s (20 °C)

Specification Test Results

About this item

 :
Boiling Pt:80…100 °C (1013 hPa)
Density:0.69 g/cm³ (20 °C)
EINECS:265-151-9
ADR:3,II
 :M-Clarity™ quality level = MQ300
Flash Pt:–10 °C
UN:1268
 :GHS02
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849