



Petroleum spirit, 60…100 °C, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:60…100 °C (1013 hPa)
Density:0.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Boiling Range:60 - 100 °C
- Residue after Evaporation:max. 0.001 %
- Water (H2O):max. 0.02 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Specification Test Results
Frequently Bought Together