Order Entry
Denmark
ContactUsLinkComponent
Petroleum spirit, 40…60 °C, PESTINORM® for pesticide residue analysis
Flammable
Health
Irritant
Environment
Petroleum spirit, 40…60 °C, PESTINORM® for pesticide residue analysis
Catalog # 83663.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
Petroleum spirit, 40…60 °C, PESTINORM® for pesticide residue analysis
Catalog # 83663.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Boiling range (40-60°C) (Vol %):
    Min. 90%
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0,02 %
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

Specification Test Results

About this item

 :
Boiling Pt:40…60 °C (1013 hPa)
Density:0.645…0.665 g/cm³ (20 °C)
EINECS:265-151-9
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–40 °C
UN:1268
REACH:01-2119474679-18
 :GHS02
 :GHS08
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849