



Petroleum spirit, 35…60 °C for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64742-49-0
:101316-46-5
MDL Number:MFCD00081849
Boiling Pt:35…60 °C
Density:0.640
Flash Pt:–49 °C (–57 °F)
Storage Temperature:Ambient
EINECS:232-453-7
UN:1268
ADR:3,I
:GHS02
:GHS08
:GHS07
:GHS09
Related Information
Frequently Bought Together