Palladium ≥99.9% (metals basis), foil, Thickness: 0.025 mm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
Density:12.02 g/cm³ (20 °C)
Molecular Weight:106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together