Order Entry
Denmark
ContactUsLinkComponent
Palladium 99.9% (trace metals basis), sponge
Irritant
Flammable
Palladium 99.9% (trace metals basis), sponge
Catalog # ACRO268150010
CAS Number:  
undefined
Irritant
Flammable
Palladium 99.9% (trace metals basis), sponge
Catalog # ACRO268150010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Grey Powder
  • Total trace metal impurities:
    ≤0.1 %

Specification Test Results

About this item

 :
 :106.420 g/mol
Merck Index:12,07121
Boiling Pt:3167 °C (1013 hPa)
Melting Pt:1555 °C
Density:12,02 g/cm³ (20 °C)
EINECS:231-115-6
UN:3089
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167