Nickel, gauze woven from wire 100 mesh, Ø 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
:58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-111-4
UN:3089
ADR:4.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together