Iridium, black ≥99.8% (metals basis), powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-88-5
:
Formula:Ir
MDL Number:MFCD00011062
:192.22 g/mol
Merck Index:12,05098
Storage Temperature:Ambient
EINECS:231-095-9
UN:3089
ADR:4.1,III
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together