Order Entry
Denmark
ContactUsLinkComponent
 
Iridium ≥99.8% (metals basis), slug, Length: 6.0 mm (0.24 in), Ø 6.0 mm (0.24 in)
Catalog # 44232.KG
CAS Number:  
Iridium ≥99.8% (metals basis), slug, Length: 6.0 mm (0.24 in), Ø 6.0 mm (0.24 in)
Catalog # 44232.KG
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Environmentally Preferable:

Specifications

About this item

CAS Number:7439-88-5
 :
Formula:Ir
MDL Number:MFCD00011062
 :192.22 g/mol
Boiling Pt:5300
Melting Pt:2450
Storage Temperature:Ambient
EINECS:231-095-9