Iridium ≥99.9% (metals basis), wire, Ø 0.125 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-88-5
:
Formula:Ir
MDL Number:MFCD00011062
Boiling Pt:5300
Molecular Weight:192.22 g/mol
Melting Pt:2450
Storage Temperature:Ambient
EINECS:231-095-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together