Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tungsten

Tungsten

Glass bottle for solids, 2,5 kg
Catalog # ELTA90220.2500
Supplier:   ELTRA
Tungsten
Catalog # ELTA90220.2500
Supplier:  ELTRA
CAS Number:  7440-33-7
Tungsten

Glass bottle for solids, 2,5 kg

CAS Number:  7440-33-7
Price
5.790,00 kr
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:7440-33-7
  • Formula:W
  • MDL Number:MFCD00011461
  • Molecular Weight:183.84 g/mol
  • Merck Index:12,09945
  • Boiling Pt:5660 °C (1013 hPa)
  • Melting Pt:3410 °C
  • Density:19 g/cm³ (20 °C)
  • EINECS:231-143-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications