Order Entry
Denmark
ContactUsLinkComponent
 
Tungsten ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41638.KS
CAS Number:  
Tungsten ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41638.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001
  • Environmentally Preferable:

Specifications

About this item

CAS Number:7440-33-7
 :
Formula:W
MDL Number:MFCD00011461
 :183.84 g/mol
Merck Index:12,09945
Boiling Pt:5660 °C (1013 hPa)
Melting Pt:3410 °C
Density:19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-143-9