Fmoc-Lys(Me)₂-OH·HCl ≥95.0% (by HPLC), Novabiochem®
Supplier: Novabiochem (Part of Merck)
Certificates
About this item
Synonyms: N-α-Fmoc-N-ε-dimethyl-N-L-lysine hydrochloride, Fmoc-Lys(Me2)-OH.HCl
CAS Number:252049-10-8
:
Formula:C₂₃H₂₉ClN₂O₄
MDL Number:MFCD05662353
:432.945 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity (IR):Passes Test
- Enantiomeric Purity:≥99.5% (a/a)
- Purity (TLC(CMA1)):≥95%
- Assay (HPLC, area%):≥95.0% (a/a)
- Solubility (1 mmole in 2 ml DMF):Clearly Soluble
- Acetate (IC):≤1.0%
Specification Test Results
Frequently Bought Together