Tungsten ≥99.95% (metals basis), plate 25x100 mm (0.98x3.9 in), Thickness: 6.4 mm (0.25 in)
Catalog # 00936.KF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Environmentally Preferable:
Specifications
About this item
:
:183.84 g/mol
Merck Index:12,09945
Boiling Pt:5660 °C (1013 hPa)
Melting Pt:3410 °C
Density:19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-143-9
CAS Number:7440-33-7
Formula:W
MDL Number:MFCD00011461