Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.8% (GC, on anhydrous basis), PESTINORM® Supra Trace
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diisopropyl ether ≥99% stabilised, TECHNICAL
Supplier: VWR Chemicals
Water, CHROMASOLV™ LC-MS Ultra tested for UHPLC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥99.8% for peptide synthesis
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Iron ≥99.5% (metals basis), spherical powder <10 µm
Supplier: Thermo Fisher Scientific