Molybdenum ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40866.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Environmentally Preferable:
Specifications
About this item
CAS Number:7439-98-7
:
Formula:Mo
MDL Number:MFCD00003465
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
:95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2