Order Entry
Denmark
ContactUsLinkComponent
Bilirubin conjugate, sodium salt
Bilirubin conjugate, sodium salt
Supplier:  Cayman Chemical
Certificates
Bilirubin conjugate, sodium salt
Supplier:  Cayman Chemical

Specifications

  • Purity:
    ≥95%
  • Molecular weight:
    842,9 g/mol
  • Storage temperature:
    –20 °C
  • Absorbance:
    λmax: 210, 452 nm
  • Molecular formula:
    C37H44N6O10S2.2Na

Specifications

Frequently Bought Together