Selenium, ingot/button, Ø ∼38 mm (1.5 in)
1
Supplier: Thermo Fisher Scientific
Selenium, ingot/button, Ø ∼38 mm (1.5 in)
Catalog # 45115.KF
Supplier: Thermo Fisher Scientific
CAS Number: 7782-49-2
Jump to:
Product Details & Documents
CAS No.: 7782-49-2 Physical Form: ≈20g/piece Odour: Odourless MDL No.: MFCD00134090
- CAS Number:7782-49-2
- Formula:Se
- MDL Number:MFCD00134090
- Molecular Weight:78.96 g/mol
- Merck Index:12,08572
- Boiling Pt:∼685 °C (1013 hPa)
- Melting Pt:217 °C
- Density:4.28 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-957-4
- UN:3283
- ADR:6.1,III
Documents
Specifications
Specifications
- Pk:1



