Order Entry
Denmark
ContactUsLinkComponent
 
Silicon ≥99.999% (metals basis), rods, Ø 0.828cm (0.326 in)
Catalog # 44625.DM
CAS Number:  
Silicon ≥99.999% (metals basis), rods, Ø 0.828cm (0.326 in)
Catalog # 44625.DM
CAS Number:  

Specifications

  • Pk:
    100 mm

Specifications

About this item

 :
 :28.09 g/mol
Merck Index:12,08635
Boiling Pt:2355 °C (1013 hPa)
Melting Pt:1410 °C
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-130-8
CAS Number:7440-21-3
Formula:Si
MDL Number:MFCD00085311