
Silicon 99+%, powder -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-21-3
:
Formula:Si
MDL Number:MFCD00085311
:28.09 g/mol
Merck Index:12,08635
Boiling Pt:2355 °C (1013 hPa)
Melting Pt:1410 °C
Density:2,34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-130-8
UN:1346
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Dark grey Powder
- Total trace metal impurities:≤1.0 %
Specification Test Results
Frequently Bought Together