Order Entry
Denmark
ContactUsLinkComponent
Graphite, plate, highly oriented pyrolytic graphite (HOPG) 10x10x1 mm (0.394x0.394x0.039 in)
Graphite, plate, highly oriented pyrolytic graphite (HOPG) 10x10x1 mm (0.394x0.394x0.039 in)
Catalog # 43836.KF
CAS Number:  
Graphite, plate, highly oriented pyrolytic graphite (HOPG) 10x10x1 mm (0.394x0.394x0.039 in)
Catalog # 43836.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
 :12.01 g/mol
Storage Temperature:Ambient
EINECS:231-955-3
CAS Number:7782-42-5
Formula:C
MDL Number:MFCD00144065