Graphite, plate, Thickness: 6.35 mm (0.250 in)
Catalog # 41831.GH
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
:12.01 g/mol
Storage Temperature:Ambient
EINECS:231-955-3
CAS Number:7782-42-5
Formula:C
MDL Number:MFCD00144065