Order Entry
Denmark
ContactUsLinkComponent
Graphite ≥99.9995% (metals basis), rods, Length: 102mm (4in), Ø 6.15mm (0.242in)
Graphite ≥99.9995% (metals basis), rods, Length: 102mm (4in), Ø 6.15mm (0.242in)
  40766.KD
 :  
Graphite ≥99.9995% (metals basis), rods, Length: 102mm (4in), Ø 6.15mm (0.242in)
  40766.KD
 :  
  • Pk:
    50
  • Seq:
    0050
 :
 :12,04560
 :3827 °C
 :2.09…2.23 g/cm³ (20 °C)
 :12.01 g/mol
 :Ambient
 :231-955-3
 :7782-42-5
 :C
 :MFCD00144065