Graphite ≥99.9995% (metals basis), rods, Length: 305mm (12in), Ø 3.05mm (0.12in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7782-42-5
:
Formula:C
MDL Number:MFCD00144065
Merck Index:12,04560
Melting Pt:3827 °C
Density:2.09…2.23 g/cm³ (20 °C)
:12.01 g/mol
Storage Temperature:Ambient
EINECS:231-955-3
Frequently Bought Together