Order Entry
Denmark
Orders LinkContactUsLinkComponent
Lead

Lead

1
Catalog # PROONIST-C2416
Health
Irritant
Environment
Lead
Catalog # PROONIST-C2416
CAS Number:  7439-92-1
CAS Number:  7439-92-1
Price
16.020,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1740 °C (1013 hPa)
  • Melting Pt:327 °C
  • Density:11.3437 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Specifications

Specifications

  • Pk:
    1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.