Order Entry
Denmark
Orders LinkContactUsLinkComponent
Precision thermometers
Supplier:   Amarell
Precision thermometers
Supplier:  Amarell
Certificates
Precision thermometers
89 Options Available Below
Product Details & Documents
These total immersion thermometers, with enclosed scale, are suitable for government testing and official calibration.
  • Milk glass scale
  • Fine subdivision up to 0,1 °C
Available with quality certificates.
 : Packaged in transparent square plastic case.
Product Family Options
With calibration certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL25990-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    −10...+30 °C
    ±0,1 %
    300 mm
    Blue special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.190,00 kr
    1.190,00 kr
    1 * 1 Items
       1.190,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25990-CB
  • AMARL25990-CB/WC
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated
    −10...+30 °C
    ±0,1 %
    300 mm
    Blue special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.470,00 kr
    2.470,00 kr
    1 * 1 Items
       2.470,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25990-CB/WC
  • AMARL25908-FL/DKD4
    Calibrated thermometer, DIN 12 778
    Calibrated thermometer, DIN 12 778
    −10...+250 °C
    ±0,1 %
    350 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    AMARL25908-FL/DKD4
    New Product
  • AMARL25926-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...150 °C
    ±0,5 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 990,00 kr
    990,00 kr
    1 * 1 Items
       990,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25926-CB
  • AMARL25944-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...50 °C
    ±0,2 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 845,00 kr
    845,00 kr
    1 * 1 Items
       845,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25944-CB
  • AMARL25944-CB/WC
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...50 °C
    ±0,2 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.560,00 kr
    1.560,00 kr
    1 * 1 Items
       1.560,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25944-CB/WC
  • AMARL25928-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...200 °C
    ±0,5 %
    400 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.080,00 kr
    1.080,00 kr
    1 * 1 Items
       1.080,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25928-CB
  • AMARL25994-FL/CC2
    Calibrated thermometer, richter-top finish with button, with capillary inside coating
    Calibrated thermometer, richter-top finish with button, with capillary inside coating
    −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.930,00 kr
    2.930,00 kr
    1 * 1 Items
       2.930,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-FL/CC2
  • AMARL25948-FL/CC3
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.690,00 kr
    2.690,00 kr
    1 * 1 Items
       2.690,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-FL/CC3
  • AMARL25992-FL/CC3
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    0...50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,0 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-FL/CC3
  • AMARL25994-FL/CC3
    Calibrated thermometer, richter-top finish with button, with capillary inside coating
    Calibrated thermometer, richter-top finish with button, with capillary inside coating
    −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.130,00 kr
    3.130,00 kr
    1 * 1 Items
       3.130,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-FL/CC3
  • AMARL25948-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.170,00 kr
    1.170,00 kr
    1 * 1 Items
       1.170,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-CB
  • AMARL25948-CB/WC
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.990,00 kr
    1.990,00 kr
    1 * 1 Items
       1.990,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-CB/WC
  • AMARL25992-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    0...50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    8,0 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.160,00 kr
    1.160,00 kr
    1 * 1 Items
       1.160,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-CB
  • AMARL25992-CB/WC
    Calibrated thermometer with works certificate, pin-top finish, capillary tube specially coated, DIN 12 775
    Calibrated thermometer with works certificate, pin-top finish, capillary tube specially coated, DIN 12 775
    0...50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    8,0 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.980,00 kr
    1.980,00 kr
    1 * 1 Items
       1.980,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-CB/WC
  • AMARL25994-CB
    Calibrated thermometer, richter-top finish with button, with capillary inside coating
    Calibrated thermometer, richter-top finish with button, with capillary inside coating
    −10...+50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.260,00 kr
    1.260,00 kr
    1 * 1 Items
       1.260,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-CB
  • AMARL25994-CB/WC
    Calibrated thermometer with works certificate, pin-top finish, with capillary inside coating
    Calibrated thermometer with works certificate, pin-top finish, with capillary inside coating
    −10...+50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.270,00 kr
    2.270,00 kr
    1 * 1 Items
       2.270,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-CB/WC
  • AMARL25998-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    0...50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.260,00 kr
    1.260,00 kr
    1 * 1 Items
       1.260,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25998-CB
  • AMARL25992-CB/DKD4
    Calibrated thermometer with DKD certificate, richter-top finish with button, capillary tube specially coated, DIN 12 775
    Calibrated thermometer with DKD certificate, richter-top finish with button, capillary tube specially coated, DIN 12 775
    0...50 °C
    ±0,1 %
    420 mm
    Blue special liquid
    8,0 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-CB/DKD4
  • AMARL25950-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    −25...+100 °C
    ±0,2 %
    450 mm
    Blue special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.360,00 kr
    1.360,00 kr
    1 * 1 Items
       1.360,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25950-CB
  • AMARL25996-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    −20...+50 °C
    ±0,1 %
    450 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.410,00 kr
    1.410,00 kr
    1 * 1 Items
       1.410,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25996-CB
  • AMARL26000-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    −10...+60 °C
    ±0,1 %
    450 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.360,00 kr
    1.360,00 kr
    1 * 1 Items
       1.360,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26000-CB
  • AMARL25952-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    −5...+150 °C
    ±0,2 %
    480 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.620,00 kr
    1.620,00 kr
    1 * 1 Items
       1.620,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25952-CB
  • AMARL25952-CB/WC
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated
    −5...+150 °C
    ±0,2 %
    480 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.730,00 kr
    2.730,00 kr
    1 * 1 Items
       2.730,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25952-CB/WC
  • AMARL26004-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    0...100 °C
    ±0,1 %
    550 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.980,00 kr
    1.980,00 kr
    1 * 1 Items
       1.980,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26004-CB
  • AMARL26004-CB/WC
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer with works certificate, richter-top finish with button, capillary tube specially coated
    0...100 °C
    ±0,1 %
    550 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.190,00 kr
    3.190,00 kr
    1 * 1 Items
       3.190,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26004-CB/WC
  • AMARL26012-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    50...150 °C
    ±0,1 %
    550 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.780,00 kr
    2.780,00 kr
    1 * 1 Items
       2.780,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26012-CB
  • AMARL26004-CB/DKD4
    Calibrated thermometer with DKD certificate, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer with DKD certificate, richter-top finish with button, capillary tube specially coated
    0...100 °C
    ±0,1 %
    550 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26004-CB/DKD4
  • AMARL26006-CB
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    Calibrated thermometer, richter-top finish with button, capillary tube specially coated
    −10...+100 °C
    ±0,1 %
    600 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.280,00 kr
    2.280,00 kr
    1 * 1 Items
       2.280,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26006-CB
With DKD certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL25902-FL/DKD3
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...100 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.040,00 kr
    2.040,00 kr
    1 * 1 Items
       2.040,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25902-FL/DKD3
  • AMARL25904-FL/DKD5
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...150 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25904-FL/DKD5
  • AMARL25902FLDKD3CL
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...100 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.040,00 kr
    2.040,00 kr
    1 * 1 Items
       2.040,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25902FLDKD3CL
  • AMARL25906-FL/DKD5
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...200 °C
    ±1,0 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25906-FL/DKD5
  • AMARL25904-FL/DKD1
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...150 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25904-FL/DKD1
  • AMARL25908-FL/DKD3
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...250 °C
    ±1,0 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25908-FL/DKD3
  • AMARL25992-FL/DKD3
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    0...50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-FL/DKD3
  • AMARL25992-FL/DKD4
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    0...50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.970,00 kr
    2.970,00 kr
    1 * 1 Items
       2.970,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-FL/DKD4
  • AMARL25948-FL/DKD5
    Richter-top finish with button, capillary lens front, DIN 12 775
    Richter-top finish with button, capillary lens front, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 3.080,00 kr
    3.080,00 kr
    1 * 1 Items
       3.080,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-FL/DKD5
  • AMARL25994-FL/DKD3
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-FL/DKD3
  • AMARL25930-FL/DKD4
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...250 °C
    ±0,5 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25930-FL/DKD4
  • AMARL25952-FL/DKD6
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    −5...+150 °C
    ±0,2 %
    480 mm
    Red special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25952-FL/DKD6
  • AMARL26006-FL/DKD3
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    −10...+100 °C
    ±0,1 %
    600 mm
    Blue special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26006-FL/DKD3
With verification certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL25922-FL/VC
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...50 °C
    ±0,5 %
    220 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.240,00 kr
    2.240,00 kr
    1 * 1 Items
       2.240,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25922-FL/VC
  • AMARL25900-FL/VC
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...50 °C
    ±1,0 %
    250 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.680,00 kr
    2.680,00 kr
    1 * 1 Items
       2.680,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25900-FL/VC
  • AMARL25902-FL/VC
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...100 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.880,00 kr
    1.880,00 kr
    1 * 1 Items
       1.880,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25902-FL/VC
  • AMARL25904-FL/VC
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...150 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.780,00 kr
    2.780,00 kr
    1 * 1 Items
       2.780,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25904-FL/VC
  • AMARL25906-FL/VC
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...200 °C
    ±1,0 %
    350 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.870,00 kr
    2.870,00 kr
    1 * 1 Items
       2.870,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25906-FL/VC
  • AMARL25910-GA/VC
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...360 °C
    ±1,0 %
    380 mm
    Gallium liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.260,00 kr
    5.260,00 kr
    1 * 1 Items
       5.260,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25910-GA/VC
  • AMARL25992-GA/VC
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    0...50 °C
    ±0,1 %
    420 mm
    Gallium liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-GA/VC
With works certificate
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL25922-FL/WC
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...50 °C
    ±0,5 %
    220 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.040,00 kr
    1.040,00 kr
    1 * 1 Items
       1.040,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25922-FL/WC
  • AMARL25900-FL/WC
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...50 °C
    ±1,0 %
    250 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.150,00 kr
    1.150,00 kr
    1 * 1 Items
       1.150,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25900-FL/WC
  • AMARL25924-FL/WC
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...100 °C
    ±0,5 %
    270 mm
    Red special liquid
    8,5 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.090,00 kr
    1.090,00 kr
    1 * 1 Items
       1.090,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25924-FL/WC
  • AMARL25902-FL/WC
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...100 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.310,00 kr
    1.310,00 kr
    1 * 1 Items
       1.310,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25902-FL/WC
  • AMARL25904-FL/WC
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...150 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.220,00 kr
    1.220,00 kr
    1 * 1 Items
       1.220,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25904-FL/WC
  • AMARL25908-FL/WC
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...250 °C
    ±1,0 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.700,00 kr
    1.700,00 kr
    1 * 1 Items
       1.700,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25908-FL/WC
  • AMARL25910-GA/WC
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...360 °C
    ±1,0 %
    380 mm
    Gallium liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25910-GA/WC
  • AMARL25948-FL/WC
    Richter-top finish with button, capillary lens front, DIN 12 775
    Richter-top finish with button, capillary lens front, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.470,00 kr
    1.470,00 kr
    1 * 1 Items
       1.470,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-FL/WC
  • AMARL25992-FL/WC
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    0...50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.460,00 kr
    1.460,00 kr
    1 * 1 Items
       1.460,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-FL/WC
  • AMARL25994-FL/WC
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.700,00 kr
    1.700,00 kr
    1 * 1 Items
       1.700,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-FL/WC
  • AMARL25944-FL/WC
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...50 °C
    ±0,2 %
    480 mm
    Red special liquid
    8,5 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.190,00 kr
    1.190,00 kr
    1 * 1 Items
       1.190,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25944-FL/WC
  • AMARL25952-FL/WC
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    −5...+150 °C
    ±0,2 %
    480 mm
    Red special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.140,00 kr
    2.140,00 kr
    1 * 1 Items
       2.140,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25952-FL/WC
  • AMARL25954-FL/WC
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    −5...+200 °C
    ±0,2 %
    550 mm
    Red special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25954-FL/WC
Product Information
  • Range
    Accuracy
    Length
    Filling
    Ø
    Availability
    Price
  • AMARL25922-FL
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...50 °C
    ±0,5 %
    220 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 385,00 kr
    385,00 kr
    1 * 1 Items
       385,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25922-FL
  • AMARL25900-FL
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...50 °C
    ±1,0 %
    250 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 323,00 kr
    323,00 kr
    1 * 1 Items
       323,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25900-FL
  • AMARL25924-FL
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...100 °C
    ±0,5 %
    270 mm
    Red special liquid
    8,5 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 436,00 kr
    436,00 kr
    1 * 1 Items
       436,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25924-FL
  • AMARL25924-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 775
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 775
    0...100 °C
    ±0,5 %
    270 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 740,00 kr
    740,00 kr
    1 * 1 Items
       740,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25924-FL-PTFE
  • AMARL25902-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 778
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 778
    0...100 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 667,00 kr
    667,00 kr
    1 * 1 Items
       667,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25902-FL-PTFE
  • AMARL25904-FL
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...150 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 385,00 kr
    385,00 kr
    1 * 1 Items
       385,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25904-FL
  • AMARL25904-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 778
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 778
    0...150 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 714,00 kr
    714,00 kr
    1 * 1 Items
       714,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25904-FL-PTFE
  • AMARL25902-FL
    Richter-top finish with button, capillary lens front, DIN 12 778
    Richter-top finish with button, capillary lens front, DIN 12 778
    0...100 °C
    ±1,0 %
    305 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 337,00 kr
    337,00 kr
    1 * 1 Items
       337,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25902-FL
  • AMARL25906-FL
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...200 °C
    ±1,0 %
    350 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 480,00 kr
    480,00 kr
    1 * 1 Items
       480,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25906-FL
  • AMARL25906-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism
    Pin-top finish, non polluting type with PTFE coated, capillary prism
    0...200 °C
    ±1,0 %
    350 mm
    Red special liquid
    8,0 - 8,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.080,00 kr
    1.080,00 kr
    1 * 1 Items
       1.080,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25906-FL-PTFE
  • AMARL25908-FL
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...250 °C
    ±1,0 %
    350 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 573,00 kr
    573,00 kr
    1 * 1 Items
       573,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25908-FL
  • AMARL25926-FL
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...150 °C
    ±0,5 %
    350 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 522,00 kr
    522,00 kr
    1 * 1 Items
       522,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25926-FL
  • AMARL25910-GA
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    0...360 °C
    ±1,0 %
    380 mm
    Gallium liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.050,00 kr
    2.050,00 kr
    1 * 1 Items
       2.050,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25910-GA
  • AMARL25912-GA
    Richter-top finish with button, capillary prism, DIN 12 778
    Richter-top finish with button, capillary prism, DIN 12 778
    −10...+410 °C
    ±1,0 %
    380 mm
    Gallium liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.180,00 kr
    2.180,00 kr
    1 * 1 Items
       2.180,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25912-GA
  • AMARL25928-FL
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...200 °C
    ±0,5 %
    400 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 595,00 kr
    595,00 kr
    1 * 1 Items
       595,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25928-FL
  • AMARL25930-FL
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...250 °C
    ±0,5 %
    420 mm
    Blue special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 697,00 kr
    697,00 kr
    1 * 1 Items
       697,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25930-FL
  • AMARL25948-FL
    Richter-top finish with button, capillary lens front, DIN 12 775
    Richter-top finish with button, capillary lens front, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 649,00 kr
    649,00 kr
    1 * 1 Items
       649,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-FL
  • AMARL25948-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 775
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 775
    0...100 °C
    ±0,2 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.070,00 kr
    1.070,00 kr
    1 * 1 Items
       1.070,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25948-FL-PTFE
  • AMARL25992-FL
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    0...50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 642,00 kr
    642,00 kr
    1 * 1 Items
       642,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-FL
  • AMARL25992-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 778
    Pin-top finish, non polluting type with PTFE coated, capillary prism, DIN 12 778
    0...50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.070,00 kr
    1.070,00 kr
    1 * 1 Items
       1.070,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25992-FL-PTFE
  • AMARL25994-FL
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 698,00 kr
    698,00 kr
    1 * 1 Items
       698,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-FL
  • AMARL25994-FL-PTFE
    Pin-top finish, non polluting type with PTFE coated, capillary prism
    Pin-top finish, non polluting type with PTFE coated, capillary prism
    −10...+50 °C
    ±0,1 %
    420 mm
    Red special liquid
    8,5 - 9,0 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.120,00 kr
    1.120,00 kr
    1 * 1 Items
       1.120,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25994-FL-PTFE
  • AMARL25952-FL
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    −5...+150 °C
    ±0,2 %
    480 mm
    Red special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.030,00 kr
    1.030,00 kr
    1 * 1 Items
       1.030,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25952-FL
  • AMARL25934-GA
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...360 °C
    ±0,5 %
    480 mm
    Gallium liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25934-GA
  • AMARL25944-FL
    Richter-top finish with button, capillary lens front
    Richter-top finish with button, capillary lens front
    0...50 °C
    ±0,2 %
    480 mm
    Red special liquid
    8,5 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 469,00 kr
    469,00 kr
    1 * 1 Items
       469,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL25944-FL
  • AMARL26004-FL
    Richter-top finish with button, capillary prism
    Richter-top finish with button, capillary prism
    0...100 °C
    ±0,1 %
    550 mm
    Red special liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1.200,00 kr
    1.200,00 kr
    1 * 1 Items
       1.200,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    AMARL26004-FL
  • AMARL25934GADKD4CL
    Richter-top finish with button, capillary prism, DIN 12 775
    Richter-top finish with button, capillary prism, DIN 12 775
    0...360 °C
    ±0,5 %
    480 mm
    Gallium liquid
    9,0 - 9,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    AMARL25934GADKD4CL
    New Product