Order Entry
Denmark
Orders LinkContactUsLinkComponent
Safety helmets, Vision Plus
Safety helmets, Vision Plus
Safety helmet, unvented, with ratchet, White
Catalog # CENUS10PLUSEWR
Safety helmets, Vision Plus
Catalog # CENUS10PLUSEWR
Safety helmets, Vision Plus
Safety helmet, unvented, with ratchet, White
Price
390,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The Vision Plus helmet offers wearers of prescription glasses total safety reassurance – with moulded integrated eye protection certified to grade A high impact levels. Helmet shell is made from ABS plastic.
  • Optical clarity to Class 1 for continuous use
  • Large side pull tabs enable the wearer to quickly and easily move the visor up and down even when wearing gloves
  • Moulded design provides improved lower cheek coverage
  • Anti-scratch and anti-mist coating for improved durability and visibility
  • Wheel ratchet, non-vented
Weight: 525 g with fitted visor
Certifications: Helmet: EN 397; Integrated visor: EN 166
Specifications
  • Colour:
    White
  • Description:
    Safety helmet, unvented, with ratchet
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Euro clips also for use on Nexus and Vision Plus helmets, S565
    Euro clips also for use on Nexus and Vision Plus helmets, S565
    CENUS565
    Log in for availability
    1 * 1 Items - 33,40 kr