Peptide A
Catalog # ENZOBMLP3060500
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:0,5 mg
- Source:
- Species:
- Seq:02
- Environmentally Preferable:
- Protein/peptide name:Peptide A
- Purity:97%
Specifications
About this item
Src kinase inhibitor
Sequence is pp60v-src (137-157), which is located in the SH3/SH2 domain region. Non-competitive inhibitor of Src and EGFR. Selectively inhibits protein tyrosine phosphorylation in cell extracts. Peptide is not phosphorylated by Src. IC50=7,5 µM for v-Src. Inhibitory target seems to be a catalytic negative regulatory domain of the tyrosine kinase.