Order Entry
Denmark
ContactUsLinkComponent
 
[pSer78]HSP27 (human), ELISA Kit
  ENZOADI900165
New Product
 :  
[pSer78]HSP27 (human), ELISA Kit
  ENZOADI900165
New Product
 :  

 

  • Pk:
    1 KIT
  • Host:
  • Primary antibody reactivity:
    Human
  • Target protein:
    [pSer78]HSP27
  • Description:
    [pSer78]HSP27 (human), ELISA kit
  • Seq:
    02
  • Environmentally Preferable:
  • Sensitivity:
    4,30 pg/ml (range 31,25 - 1000 pg/ml)

 

 

Most comprehensive portfolio of heat shock protein (chaperone) products commercially available.

For the quantitative determination of human [pSer78]HSP27 in cell lysates, plasma, and serum.

The [pSer78]HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.

 : Contains Microtiter plate, Conjugate, Antibody, Assay buffer 27, Wash buffer concentrate, Standard, TMB Substrate, 5X Extraction reagent, Stop solution 2.